
2,6-Dimethylhydroquinone ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:654-42-2
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00016466
:138.17 g/mol
EINECS:211-505-2
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to Light yellow to Light red powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Cerium redox Method):min. 98.0 %
- Melting point:151.0 to 155.0 deg-C
Specification Test Results
Frequently Bought Together