
5-Chloro-2-mercaptobenzothiazole ≥96.0% (by titrimetric analysis)
Catalog # TCIAC1297-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Yellow to Green powder to crystal
- Purity (Neutralization titration):min. 96.0 %
- Melting point:198.0 to 204.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
About this item
:
:201.7 g/mol
EINECS:226-235-0
:GHS07
CAS Number:5331-91-9
Formula:C₇H₄ClNS₂
MDL Number:MFCD00005783