Order Entry
Netherlands
ContactUsLinkComponent
Tetraoctylphosphonium bromide ≥98.0% (by titrimetric analysis)
Irritant
Tetraoctylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1650-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Tetraoctylphosphonium bromide ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1650-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to lump
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    42.0 to 46.0 deg-C

Specification Test Results

About this item

 :
 :563.77 g/mol
EINECS:245-935-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:23906-97-0
Formula:C₃₂H₆₈BrP
MDL Number:MFCD00210101