2-Bromoadamantane ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:7314-85-4
:
Formula:C₁₀H₁₅Br
MDL Number:MFCD00074745
:215.13 g/mol
Boiling Pt:230 °C (1013 hPa)
Melting Pt:138…140 °C
EINECS:230-777-3
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together