Order Entry
Netherlands
ContactUsLinkComponent
2,6-Di-tert-butyl-4-methylpyridine ≥98.0% (by GC, titration analysis)
Irritant
2,6-Di-tert-butyl-4-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
Irritant
2,6-Di-tert-butyl-4-methylpyridine ≥98.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to lump
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

Frequently Bought Together