
2-(Methylamino)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:4597-87-9
:
Formula:C₆H₈N₂
MDL Number:MFCD00006250
:108.14 g/mol
Boiling Pt:201°C
Melting Pt:15 °C
Density:1,0707 g/cm³ (17.7 °C)
Flash Pt:102°C
EINECS:224-997-9
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together