
2,6-Diaminopyridine ≥98.0% (by GC, titration analysis)
Catalog # TCIAD1154-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:120.0 to 124.0 deg-C
Specification Test Results
About this item
:
:109.13 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:115…118 °C
Flash Pt:155 °C
Storage Temperature:Ambient
EINECS:205-507-2
ADR:6.1,II
:GHS07
CAS Number:141-86-6
Formula:C₅H₇N₃
MDL Number:MFCD00006329