Order Entry
Netherlands
ContactUsLinkComponent
2,6-Diisopropylnaphthalene ≥99.0% (by GC)
2,6-Diisopropylnaphthalene ≥99.0% (by GC)
Catalog # TCIAD1598-25G
Supplier:  TCI
CAS Number:  
undefined
2,6-Diisopropylnaphthalene ≥99.0% (by GC)
Catalog # TCIAD1598-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    69.0 to 71.0 deg-C
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :212.33 g/mol
Boiling Pt:279,3 °C (1013 hPa)
Melting Pt:67…69 °C
Density:0,95 g/cm³ (20 °C)
EINECS:246-045-1
CAS Number:24157-81-1
Formula:C₁₆H₂₀
MDL Number:MFCD00021648