
1,4-Dibromonaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:83-53-4
:
Formula:C₁₀H₆Br₂
MDL Number:MFCD00041823
:285.97 g/mol
Boiling Pt:310 °C (1013 hPa)
Melting Pt:92 °C
EINECS:201-484-8
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:81.0 to 84.0 deg-C
- Solubility in hot Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together