
(S)-(-)-N-(1-Phenylethyl)phthalamic acid ≥98.0% (by titrimetric analysis)
Catalog # TCIAM0824-1G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-44.0 to -48.0 deg (C=2, EtOH)
- Solubility in Methanol:very faint turbidity
Specification Test Results
About this item
:
:269.3
EINECS:244-571-6
:GHS07
CAS Number:21752-36-3
Formula:C₁₆H₁₅NO₃
MDL Number:MFCD00013979