
3-Hydrazinobenzoic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:38235-71-1
:
Formula:C₇H₈N₂O₂
MDL Number:MFCD00191447
:152.15 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together