Order Entry
Netherlands
ContactUsLinkComponent
Girard's reagent D (N,N-dimethylglycine hydrazide dihydrochloride) ≥99.0% (by titrimetric analysis)
Girard's reagent D (N,N-dimethylglycine hydrazide dihydrochloride) ≥99.0% (by titrimetric analysis)
Catalog # TCIAD0727-10G
Supplier:  TCI
CAS Number:  
undefined
Girard's reagent D (N,N-dimethylglycine hydrazide dihydrochloride) ≥99.0% (by titrimetric analysis)
Catalog # TCIAD0727-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Bromination Method):
    min. 99.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :190.07 g/mol
Merck Index:14,03245
EINECS:208-721-4
CAS Number:5787-71-3
 :MFCD06656061
Formula:C₄H₁₃Cl₂N₃O
MDL Number:MFCD00042020