
Butaandioldiglycidylether ≥93.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2425-79-8
:
Formula:C₁₀H₁₈O₄
MDL Number:MFCD00005146
:202.25 g/mol
Boiling Pt:160°C
Flash Pt:140°C
EINECS:219-371-7
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 93.0 %
Specification Test Results
Frequently Bought Together