Order Entry
Netherlands
ContactUsLinkComponent
 
 
 
1,3-Diiodo-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
 
1,3-Diiodo-5,5-dimethylhydantoin ≥97.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Iodometric Titration):
    min. 97.0 %

 

Frequently Bought Together