


N,N-Dimethyl-1,3-diaminopropane ≥99.0% (by GC, titration analysis)
: TCI
:109-55-7
:
:C₅H₁₄N₂
:MFCD00008216
:102.18 g/mol
:136°C
:38°C
:203-680-9
:2734
:8,II
:GHS02
:GHS05
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 99.0 %
- Purity (Nonaqueous Titration):min. 99.0 %
Frequently Bought Together