Order Entry
Netherlands
ContactUsLinkComponent
Dibutyl phosphonate ≥95.0% (by GC)
Irritant
Dibutyl phosphonate ≥95.0% (by GC)
Catalog # TCIAD0300-25G
Supplier:  TCI
CAS Number:  
Irritant
Dibutyl phosphonate ≥95.0% (by GC)
Catalog # TCIAD0300-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :194.21 g/mol
Boiling Pt:121°C
Flash Pt:123°C
EINECS:217-316-1
 :GHS07
CAS Number:1809-19-4
Formula:C₈H₁₉O₃P
MDL Number:MFCD00066633