Order Entry
Netherlands
ContactUsLinkComponent
 
Dibutyl succinate ≥99.0% (by GC)
  TCIAS0102-25ML
 :  TCI
 :  
undefined
Dibutyl succinate ≥99.0% (by GC)
  TCIAS0102-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

 

 

 :
 :230.3 g/mol
 :274°C
 :-29°C
 :–29°C
 :144°C
 :205-449-8
 :141-03-7
 :C₁₂H₂₂O₄
 :MFCD00040495