

2-Bromoethyl ethyl ether ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:592-55-2
:
Formula:C₄H₉BrO
MDL Number:MFCD00000237
:153.02 g/mol
Flash Pt:21°C
Storage Temperature:Freezer
EINECS:209-763-6
UN:2340
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together