Order Entry
Netherlands
ContactUsLinkComponent
Sodium azide ≥99.0% (by titrimetric analysis)
Corrosive
Toxic
Health
Environment
Sodium azide ≥99.0% (by titrimetric analysis)
Catalog # TCIAS0489-100G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
Health
Environment
Sodium azide ≥99.0% (by titrimetric analysis)
Catalog # TCIAS0489-100G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Cerium redox Method):
    min. 99.0 %

Specification Test Results

About this item

 :
 :65.01 g/mol
Merck Index:14,08581
Melting Pt:275 °C
Density:1,85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:247-852-1
UN:1687
ADR:6.1,II
 :GHS05
 :GHS06
 :GHS08
 :GHS09
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536