

cis-1,3-Dimethylcyclohexane ≥99.0% (by GC)
: TCI
:638-04-0
:
:C₈H₁₆
:MFCD00001507
:112.22 g/mol
:120°C
:–75.5 °C
:0,766 g/cm³ (20 °C)
:6°C
:211-316-5
:2263
:3,II
:GHS02
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together