Order Entry
Netherlands
ContactUsLinkComponent
 
 
cis-3,3,5-Trimethylcyclohexanol ≥96.0% (by GC)
  TCIAT0874-25G
 :  TCI
 :  
undefined
 
cis-3,3,5-Trimethylcyclohexanol ≥96.0% (by GC)
  TCIAT0874-25G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

 

 

  • Appearance:
    White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :142.24 g/mol
 :196,5 °C (1013 hPa)
 :53…54 °C
 :0,86 g/cm³ (20 °C)
 :213-268-0
 :GHS05
 :933-48-2
 :C₉H₁₈O
 :MFCD00070479