2-Ethyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
TCIAE0507-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:174.0 to 177.0 deg-C
- Solubility in Methanol:almost transparency
:
:126.16 g/mol
:205…210 °C
:Refrigerator
:212-512-3
:823-36-9
:C₇H₁₀O₂
:MFCD00044191