
2,6-Di-tert-butyl-4-methylpyrylium trifluoromethanesulphonate ≥97.0% (by titrimetric analysis)
TCIAD2420-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:170.0 to 173.0 deg-C
:
:356.41 g/mol
:GHS07
:59643-43-5
:C₁₅H₂₃F₃O₄S
:MFCD00229854