

Methyl-3-(trifluoromethyl)benzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2557-13-3
:
Formula:C₉H₇F₃O₂
MDL Number:MFCD00043543
:204.15 g/mol
Boiling Pt:198°C
Flash Pt:40°C
UN:3272
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together