
Cerium(III) trifluoromethanesulphonate ≥90.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:76089-77-5
:
Formula:C₃CeF₉O₉S₃
MDL Number:MFCD02093899
ADR:8,III
:587.33 g/mol
UN:1759
:GHS05
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Chelometric Titration):min. 95.0 % (calcd.on anh.substance)
- Water:max. 7.0 %
Specification Test Results
Frequently Bought Together