Order Entry
Netherlands
ContactUsLinkComponent
 
 
Magnesium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
Magnesium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Chelometric Titration):
    min. 98.0 %

 

Frequently Bought Together