Order Entry
Netherlands
ContactUsLinkComponent
1,1'-Bis(diphenylphosphino)ferrocene ≥96.0% (by titrimetric analysis)
Irritant
1,1'-Bis(diphenylphosphino)ferrocene ≥96.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1,1'-Bis(diphenylphosphino)ferrocene ≥96.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystaline
  • Purity (Chelometric Titration):
    min. 96.0 %

Specification Test Results

Frequently Bought Together