

Chloro(diisopropyl)phosphine ≥98.0% (by titrimetric analysis)
Catalog # TCIAC1720-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
About this item
:
:152.6 g/mol
Boiling Pt:158°C
Flash Pt:4°C
Storage Temperature:Refrigerator
UN:2924
ADR:3,II
:GHS02
:GHS05
CAS Number:40244-90-4
Formula:C₆H₁₄ClP
MDL Number:MFCD00015027