Order Entry
Netherlands
ContactUsLinkComponent
Methyl-2,4-difluorobenzoate ≥98.0% (by GC)
Corrosive
Irritant
Methyl-2,4-difluorobenzoate ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
Methyl-2,4-difluorobenzoate ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together