

Diethylene glycol monohexyl ether ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:112-59-4
:
Formula:C₁₀H₂₂O₃
MDL Number:MFCD00010703
:190.28 g/mol
Flash Pt:141°C
EINECS:203-988-3
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together