
Copper(II) trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:34946-82-2
:
Formula:C₂CuF₆O₆S₂
MDL Number:MFCD00077492
:361.69 g/mol
EINECS:252-300-8
UN:1759
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Gray to Dark blue powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together