Order Entry
Netherlands
ContactUsLinkComponent
Potassium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Irritant
Potassium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1549-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Potassium trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Catalog # TCIAT1549-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Ion exchange titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :188.17 g/mol
 :GHS07
CAS Number:2926-27-4
Formula:CF₃KO₃S
MDL Number:MFCD00042370