
1,3-Cyclohexanedione ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:504-02-9
:
Formula:C₆H₈O₂
MDL Number:MFCD00001585
:112.13 g/mol
Merck Index:14,03178
Boiling Pt:235 °C (1013 hPa)
Melting Pt:105 °C
Density:1,25 g/cm³ (20 °C)
EINECS:207-980-0
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:103.0 to 106.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together