
4-Methylquinoline ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:491-35-0
:
Formula:C₁₀H₉N
MDL Number:MFCD00006784
:143.19 g/mol
Merck Index:14,05441
Boiling Pt:265°C
Melting Pt:9 °C
Density:1,0995 g/cm³ (0 °C)
EINECS:207-734-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Yellow to Orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 97.0 %
Specification Test Results
Frequently Bought Together