

2-Methyl-8-quinolinol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:826-81-3
:
Formula:C₁₀H₉NO
MDL Number:MFCD00006765
:159.19 g/mol
Boiling Pt:145°C
Melting Pt:70…72 °C
Storage Temperature:Ambient
EINECS:212-562-6
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:72.0 to 74.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together