


Quinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:91-22-5
:
Formula:C₉H₇N
MDL Number:MFCD00006736
:129.16 g/mol
Merck Index:14,08068
Boiling Pt:238°C
Melting Pt:–15°C
Density:1,095 g/cm³ (20 °C)
Flash Pt:106°C (closed cup)
Storage Temperature:Ambient
EINECS:202-051-6
UN:2656
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together