Order Entry
Netherlands
ContactUsLinkComponent
2,3,5-Trimethylanisole ≥97.0% (by GC)
2,3,5-Trimethylanisole ≥97.0% (by GC)
Catalog # TCIAT2456-5G
Supplier:  TCI
CAS Number:  
undefined
2,3,5-Trimethylanisole ≥97.0% (by GC)
Catalog # TCIAT2456-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :150.22 g/mol
Boiling Pt:216°C
EINECS:243-843-1
CAS Number:20469-61-8
Formula:C₁₀H₁₄O
MDL Number:MFCD00142783