Order Entry
Netherlands
ContactUsLinkComponent
2,3-Dimethylaniline ≥99.0% (by GC, titration analysis)
Toxic
Health
Environment
2,3-Dimethylaniline ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Toxic
Health
Environment
2,3-Dimethylaniline ≥99.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (GC):
    min. 99.0 %
  • Purity (Nonaqueous Titration):
    min. 99.0 %

Specification Test Results

Frequently Bought Together