Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-m-xylene ≥98.0% (by GC)
Irritant
2-Chloro-m-xylene ≥98.0% (by GC)
Catalog # TCIAC2164-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2-Chloro-m-xylene ≥98.0% (by GC)
Catalog # TCIAC2164-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Boiling Pt:187°C
Melting Pt:-36°C
Density:1,053 g/cm³ (20 °C)
Flash Pt:63°C
 :140.61 g/mol
EINECS:229-843-4
 :GHS07
CAS Number:6781-98-2
Formula:C₈H₉Cl
MDL Number:MFCD00000565
Melting Pt:–36°C