
3-Bromothiophene-2-carbaldehyde ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:930-96-1
:
Formula:C₅H₃BrOS
MDL Number:MFCD00126680
:191.05 g/mol
Boiling Pt:115°C
:GHS07
Specification Test Results
- Appearance:Light yellow to Amber to Dark green clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
Frequently Bought Together