Order Entry
Netherlands
ContactUsLinkComponent
N-(3-Hydroxypropyl)morpholine ≥98.0% (by GC)
Irritant
N-(3-Hydroxypropyl)morpholine ≥98.0% (by GC)
Catalog # TCIAH1355-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
N-(3-Hydroxypropyl)morpholine ≥98.0% (by GC)
Catalog # TCIAH1355-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :145.2 g/mol
Boiling Pt:241°C
Flash Pt:85°C
 :GHS07
CAS Number:4441-30-9
Formula:C₇H₁₅NO₂
MDL Number:MFCD01713533