Order Entry
Netherlands
ContactUsLinkComponent
N-(3-Aminopropyl)morpholine ≥99.0% (by GC)
Corrosive
Irritant
N-(3-Aminopropyl)morpholine ≥99.0% (by GC)
Supplier:  TCI
Certificates
undefined
Corrosive
Irritant
N-(3-Aminopropyl)morpholine ≥99.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

Related Information

Frequently Bought Together