
3-Fluorophenethyl bromide ≥97.0% (by GC)
TCIAB2159-5G
: TCI
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 97.0 %
:
:203.05 g/mol
:94°C
:GHS07
:25017-13-4
:C₈H₈BrF
:MFCD03095397