

3,4-Dimethoxyphenethylamine ≥98.0% (by GC, titration analysis)
: TCI
:120-20-7
:
:C₁₀H₁₅NO₂
:MFCD00008188
:181.23
:130°C
:204-376-9
:2735
:8,III
:GHS05
:GHS07
- Appearance:Colorless to Red to Green clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Frequently Bought Together