Order Entry
Netherlands
ContactUsLinkComponent
 
 
2,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
  TCIAD2925-5G
 :  TCI
 :  
undefined
 
2,4-Dichlorophenethylamine ≥98.0% (by GC, titration analysis)
  TCIAD2925-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :190.07 g/mol
 :GHS07
 :52516-13-9
 :C₈H₉Cl₂N
 :MFCD00060611