Order Entry
Netherlands
ContactUsLinkComponent
α'-Bromo-α,α,α-trifluoro-o-xylene ≥97.0% (by GC, titration analysis)
Corrosive
α'-Bromo-α,α,α-trifluoro-o-xylene ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Corrosive
α'-Bromo-α,α,α-trifluoro-o-xylene ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %

Specification Test Results

Related Information

Frequently Bought Together