Order Entry
Netherlands
ContactUsLinkComponent
α-Bromo-p-xylene ≥98.0% (by GC)
Corrosive
Toxic
α-Bromo-p-xylene ≥98.0% (by GC)
Catalog # TCIAB0138-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
α-Bromo-p-xylene ≥98.0% (by GC)
Catalog # TCIAB0138-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White or Colorless to Light yellow powder to lump to clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
Merck Index:14,10090
Boiling Pt:218 °C (1013 hPa)
Melting Pt:38 °C
Density:1,324 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:203-240-6
UN:3417
ADR:6.1,II
 :185.06 g/mol
 :GHS05
 :GHS06
CAS Number:104-81-4
Formula:C₈H₉Br
MDL Number:MFCD00000181