Order Entry
Netherlands
ContactUsLinkComponent
α,α'-Dibromo-p-xylene ≥98.0% (by GC)
Corrosive
Toxic
α,α'-Dibromo-p-xylene ≥98.0% (by GC)
Catalog # TCIAD0216-25G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
Toxic
α,α'-Dibromo-p-xylene ≥98.0% (by GC)
Catalog # TCIAD0216-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    144.0 to 147.0 deg-C
  • Solubility in hot Toluene:
    almost transparency

Specification Test Results

About this item

 :
 :263.96 g/mol
Boiling Pt:245 °C (1013 hPa)
Density:2,012 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:210-781-1
UN:2928
ADR:6.1,II
 :GHS05
 :GHS06
CAS Number:623-24-5
Formula:C₈H₈Br₂
MDL Number:MFCD00000182