
4-tert-Butylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:769-92-6
:
Formula:C₁₀H₁₅N
MDL Number:MFCD00007899
Boiling Pt:230°C
Melting Pt:15 °C
Density:0,941 g/cm³ (20 °C)
Flash Pt:101°C
Storage Temperature:Refrigerator
EINECS:212-215-9
ADR:8,III
:149.24 g/mol
:GHS07
Specification Test Results
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together