
α,α,α-Trifluoro-3-nitrotoluene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:98-46-4
:
Formula:C₇H₄F₃NO₂
MDL Number:MFCD00007260
:191.11 g/mol
Boiling Pt:203°C
Melting Pt:–2.4 °C
Density:1,437 g/cm³ (20 °C)
Flash Pt:103°C
EINECS:202-670-1
UN:2306
ADR:6.1,II
:GHS06
Specification Test Results
- Appearance:Colorless to Yellow to Green clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
Frequently Bought Together